ContactUsLinkComponent
Human CD44 (CD44 antigen) ELISA Kit for detecting CD44 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biol ogical fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. For 1 * 96 Tests
  MSPPEKF57197
Human CD44 (CD44 antigen) ELISA Kit for detecting CD44 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biol ogical fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. For 1 * 96 Tests
  MSPPEKF57197
Frequently Bought Together