Porcine TSH (Thyroid Stimulating Hormone) ELISA Kit for detecting TSH in serum, plasma, cell culture supernatant, cell or tissue lys ate, other biological fluids. Species Reactivity: Pig (Sus scrofa Porcine). Detection Range: 1.25-80ng/ml. Sensitivi 1 * 96 Tests
Catalog # MSPPEKF60817
Supplier: MARKET SOURCE PART PROCESS