Rat IgE (Immunoglobulin E) ELISA Kit for detecting IgE in serum, plasma, cell culture supernatant, cell or tissue lysate, other biol ogical fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 3.125-200ng/ml. Sensitivity: 1.875ng/ml. 1 * 96 Tests
Catalog # MSPPEKF58258
Supplier: MARKET SOURCE PART PROCESS
Frequently Bought Together