Human KL (Klotho) ELISA Kit for detecting KL in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological flu ids. Species Reactivity: Human (Homo sapiens). Detection Range: 7.813-500pg/ml. Sensitivity: 4.688pg/ml. Format: 96-W 1 * 96 Tests
Catalog # MSPPEKF57475
Supplier: MARKET SOURCE PART PROCESS