ContactUsLinkComponent
Human SAKL (Soluble alpha-Klotho) ELISA Kit for detecting SAKL in serum, plasma, cell culture supernatant, cell or tissue lysate, ot her biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 93.75-6000pg/ml. Sensitivity: 56.25 1 * 96 Tests
  MSPPEKF58413
Human SAKL (Soluble alpha-Klotho) ELISA Kit for detecting SAKL in serum, plasma, cell culture supernatant, cell or tissue lysate, ot her biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 93.75-6000pg/ml. Sensitivity: 56.25 1 * 96 Tests
  MSPPEKF58413