Hamster CXCL10/IP-10 (Interferon Gamma Induced Protein 10kDa) ELISA Kit for detecting CXCL10 in serum, plasma, cell culture supernat ant, cell or tissue lysate, other biological fluids. Species Reactivity: Hamster. Detection Range: 15.625-1000pg/ml. 1 * 96 Tests
Catalog # MSPPEKF60825
Supplier: MARKET SOURCE PART PROCESS