Human βEP (Beta-endorphin) ELISA Kit for detecting βEP in serum, plasma, cell culture supernatant, cell or tissue lysate, other biol ogical fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 15.625-1000pg/ml. Sensitivity: 9.375pg/ml. 1 * 96 Tests
Catalog # MSPPEKF57209
Supplier: MARKET SOURCE PART PROCESS
Frequently Bought Together