ContactUsLinkComponent
Human EDN1 (Endothelin-1) ELISA Kit for detecting EDN1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biol ogical fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 1.25-80pg/ml. Sensitivity: 0.75pg/ml. Forma 1 * 96 Tests
  MSPPEKF57193
Human EDN1 (Endothelin-1) ELISA Kit for detecting EDN1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biol ogical fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 1.25-80pg/ml. Sensitivity: 0.75pg/ml. Forma 1 * 96 Tests
  MSPPEKF57193
Frequently Bought Together