Human F8 (Coagulation factor VIII) ELISA Kit for detecting F8 in plasma, cell culture supernatant, cell or tissue lysate, other biol ogical fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. For 1 * 96 Tests
Catalog # MSPPEKF60421
Supplier: MARKET SOURCE PART PROCESS
Frequently Bought Together