ContactUsLinkComponent
Bovine serum albumin (BSA) ELISA Kit for detecting BSA in serum, plasma, cell culture supernatant, cell or tissue lysate, other biol ogical fluids. Species Reactivity: Bovine (Bos taurus). Detection Range: 0.156-10ug/ml. Sensitivity: 0.094ug/ml. Form 1 * 96 Tests
  MSPPEKF60803
Bovine serum albumin (BSA) ELISA Kit for detecting BSA in serum, plasma, cell culture supernatant, cell or tissue lysate, other biol ogical fluids. Species Reactivity: Bovine (Bos taurus). Detection Range: 0.156-10ug/ml. Sensitivity: 0.094ug/ml. Form 1 * 96 Tests
  MSPPEKF60803
Frequently Bought Together