Chicken LZM (Lysozyme LZM) ELISA Kit for detecting LZM in serum, plasma, cell culture supernatant, cell or tissue lysate, other biol ogical fluids. Species Reactivity: Chicken (Gallus). Detection Range: 31.25-2000pg/ml. Sensitivity: 18.75pg/ml. Forma 1 * 96 Tests
Catalog # MSPPEKF60852
Supplier: MARKET SOURCE PART PROCESS
Frequently Bought Together