4-HNE (4-Hydroxynonenal) ELISA Kit for detecting 4-HNE in serum, plasma, cell culture supernatant, cell or tissue lysate, other biol ogical fluids. Species Reactivity: Universal. Detection Range: 31.25-2000pg/ml. Sensitivity: 18.75pg/ml. Format: 96-W 1 * 96 Tests
Catalog # MSPPEKF60360
Supplier: MARKET SOURCE PART PROCESS
Frequently Bought Together