ContactUsLinkComponent
Mouse D-Lactate Dehydrogenase, D-Ldh Elisa Kit for detecting Lactate Dehydrogenase D in serum, plasma, cell culture supernates, tiss ue homogenates. Species Reactivity: Mouse (Mus musculus). Detection Range: 4.69-300mU/ml. Sensitivity: 1.17mU/ml. For 1 * 96 Tests
  MSPPEKC36761
Mouse D-Lactate Dehydrogenase, D-Ldh Elisa Kit for detecting Lactate Dehydrogenase D in serum, plasma, cell culture supernates, tiss ue homogenates. Species Reactivity: Mouse (Mus musculus). Detection Range: 4.69-300mU/ml. Sensitivity: 1.17mU/ml. For 1 * 96 Tests
  MSPPEKC36761