ContactUsLinkComponent
Rat L-Lactate Dehydrogenase, L-Ldh Elisa Kit for detecting Lactate Dehydrogenase A in serum, plasma, tissue homogenates. Species Rea ctivity: Rat (Rattus norvegicus). Detection Range: 39-2500mU/ml. Sensitivity: 9.77mU/ml. Format: 96-Well Plate. Assay 1 * 96 Tests
  MSPPEKC39350
Rat L-Lactate Dehydrogenase, L-Ldh Elisa Kit for detecting Lactate Dehydrogenase A in serum, plasma, tissue homogenates. Species Rea ctivity: Rat (Rattus norvegicus). Detection Range: 39-2500mU/ml. Sensitivity: 9.77mU/ml. Format: 96-Well Plate. Assay 1 * 96 Tests
  MSPPEKC39350