Rat Anti-Thyroid-Peroxidase Antibody, Tpo-Ab Elisa Kit for detecting Anti-Thyroid-Peroxidase Antibody, Tpo-Ab in serum, plasma. Spec ies Reactivity: Rat (Rattus norvegicus). Detection Range: 5-80IU/ml. Sensitivity: 5IU/ml. Format: 96-Well Plate. Assa 1 * 96 Tests
Catalog # MSPPEKC38687
Supplier: MARKET SOURCE PART PROCESS
Frequently Bought Together