Mouse Anti-Thyroid-Globulin Antibody, Tgab Elisa Kit for detecting Anti-Thyroid-Globulin Antibody, Tgab in serum, plasma. Species Re activity: Mouse (Mus musculus). Detection Range: 1.25-20U/ml. Sensitivity: 1.25u/ml. Format: 96-Well Plate. Assay Typ 1 * 96 Tests
Catalog # MSPPEKC36345
Supplier: MARKET SOURCE PART PROCESS