Human Perforin-1 (Prf1/Pfp) Elisa Kit for detecting Perforin 1 (Pore Forming Protein) in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.078ng/ml. Format: 96-Well Plate. Assa 1 * 96 Tests
Catalog # MSPPEKC34955
Supplier: MARKET SOURCE PART PROCESS