Human Antidiuretic Hormone/Vasopressin/Arginine Vasopressin, Adh/Vp/Avp Elisa Kit for detecting Arginine Vasopressin in serum, plasm a, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 1.25-80pg/ml. Sensitivity: 0.312pg/ 1 * 96 Tests
Catalog # MSPPEKC32656
Supplier: MARKET SOURCE PART PROCESS