Human Mammalian Target Of Rapamyein, Mtor Elisa Kit for detecting Mammalian Target Of Rapamyein, Mtor in serum, plasma, tissue homog enates. Species Reactivity: Human (Homo sapiens). Detection Range: 1.56-100pg/ml. Sensitivity: 0.39pg/ml. Format: 96- 1 * 96 Tests
Catalog # MSPPEKC34534
Supplier: MARKET SOURCE PART PROCESS