Human Secretory Component, Sc Elisa Kit for detecting Secretory Component, Sc in serum, plasma, urine, tissue homogenates. Species R eactivity: Human (Homo sapiens). Detection Range: 0.312-20ug/ml. Sensitivity: 0.078ug/mL. Format: 96-Well Plate. Assa 1 * 96 Tests
Catalog # MSPPEKC35479
Supplier: MARKET SOURCE PART PROCESS