Mouse Kidney Injury Molecule 1, Kim-1 Elisa Kit for detecting Hepatitis A Virus Cellular Receptor 1 in serum, plasma, tissue homogen ates, urine. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.9-60ng/ml. Sensitivity: 0.225ng/ml. Format: 1 * 96 Tests
Catalog # MSPPEKC37245
Supplier: MARKET SOURCE PART PROCESS