Rat Kidney Injury Molecule 1, Kim-1 Elisa Kit for detecting Hepatitis A Virus Cellular Receptor 1 in serum, plasma, urine. Species R eactivity: Rat (Rattus norvegicus). Detection Range: 0.313-20ng/ml. Sensitivity: 0.228ng/ml. Format: 96-Well Plate. A 1 * 96 Tests
Catalog # MSPPEKC40521
Supplier: MARKET SOURCE PART PROCESS