Human Kidney Injury Molecule 1, Kim-1 Elisa Kit for detecting Hepatitis A Virus Cellular Receptor 1 in serum, urine, tissue homogena tes. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.043ng/ml. Format: 96-We 1 * 96 Tests
Catalog # MSPPEKC40520
Supplier: MARKET SOURCE PART PROCESS