Rat Nuclear Factor-Κb P65, Nf-Κb P65 Elisa Kit for detecting Nuclear Factor-Κb P65, Nf-Κb P65 in serum, plasma, tissue homogenates. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 1.56-100pg/ml. Sensitivity: 0.39pg/ml. Format: 96-Well P 1 * 96 Tests
Catalog # MSPPEKC39500
Supplier: MARKET SOURCE PART PROCESS