Rat Epinephrine/Adrenaline, Epi Elisa Kit for detecting Epinephrine/Adrenaline, Epi in serum, plasma, tissue homogenates. Species Re activity: Rat (Rattus norvegicus). Detection Range: 1-200pg/ml. Sensitivity: 0.5pg/ml. Format: 96-Well Plate. Assay T 1 * 96 Tests
Catalog # MSPPEKC39002
Supplier: MARKET SOURCE PART PROCESS