Human Estradiol Receptor, Er Elisa Kit for detecting Estrogen Receptor 1 in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 3.12-200pg/ml. Sensitivity: 0.78pg/ml. Format: 96-Well Plate. Ass 1 * 96 Tests
Catalog # MSPPEKC33633
Supplier: MARKET SOURCE PART PROCESS