Human Angiotensinogen (Agt) Elisa Kit for detecting Angiotensinogen (Serpin Peptidase Inhibitor, Clade A, Member 8) in serum, plasma , urine, cell culture supernates. Species Reactivity: Human (Homo sapiens). Detection Range: 78-5000pg/ml. Sensitivit 1 * 96 Tests
Catalog # MSPPEKC32593
Supplier: MARKET SOURCE PART PROCESS