Human Basic Fibroblast Growth Factor, Bfgf Elisa Kit for detecting Basic Fibroblast Growth Factor, Bfgf in serum, plasma, tissue hom ogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 1.56-100pg/ml. Sensitivity: 0.39pg/ml. Format: 9 1 * 96 Tests
Catalog # MSPPEKC32831
Supplier: MARKET SOURCE PART PROCESS