Rat Interleukin 12, Il-12/P70 Elisa Kit for detecting Interleukin 12, Il-12/P70 in serum, plasma, tissue homogenates. Species Reacti vity: Rat (Rattus norvegicus). Detection Range: 3.12-200pg/ml. Sensitivity: 0.78pg/ml. Format: 96-Well Plate. Assay T 1 * 96 Tests
Catalog # MSPPEKC39254
Supplier: MARKET SOURCE PART PROCESS