Mouse Stem Cell Factor/Mast Cell Growth Factor, Scf/Mgf Elisa Kit for detecting Kit Ligand in serum, plasma, tissue homogenates. Spe cies Reactivity: Mouse (Mus musculus). Detection Range: 3.12-200pg/ml. Sensitivity: 0.78pg/ml. Format: 96-Well Plate. 1 * 96 Tests
Catalog # MSPPEKC40112
Supplier: MARKET SOURCE PART PROCESS
Frequently Bought Together