Human Forkhead Box P3 (FOXP3) ELISA Kit for detecting FOXP3 in tissue homogenates, cell lysates, cell culture supernates, other biol ogical fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.117ng/ml. For 1 * 96 Tests
Catalog # MSPPEKU04262
Supplier: MARKET SOURCE PART PROCESS
Frequently Bought Together