Human Oxidized Low Density Lipoprotein (OxLDL) ELISA Kit for detecting OxLDL in serum, plasma, other biological fluids. Species Reac tivity: Human (Homo sapiens). Detection Range: 15.6-1,000pg/ml. Sensitivity: 6.4pg/ml. Format: 96-Well Plate. Assay T 1 * 96 Tests
Catalog # MSPPEKU06444
Supplier: MARKET SOURCE PART PROCESS