Human Bone Morphogenetic Protein 10 (BMP10) ELISA Kit for detecting BMP10 in serum, plasma, tissue homogenates, other biological flu ids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.056ng/ml. Format: 96-We 1 * 96 Tests
Catalog # MSPPEKU02740
Supplier: MARKET SOURCE PART PROCESS