Human Creatine Kinase, Muscle (CKM) ELISA Kit for detecting CKM in serum, plasma, tissue homogenates, cell lysates, cell culture sup ernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 1.56-100ng/ml. Sensitivi 1 * 96 Tests
Catalog # MSPPEKU03492
Supplier: MARKET SOURCE PART PROCESS