Bacitracin (Bac) ELISA Kit for detecting Bac in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biol ogical fluids. Species Reactivity: Universal. Detection Range: 8-5,000pg/ml. Sensitivity: 4pg/ml. Format: 96-Well Pla 1 * 96 Tests
Catalog # MSPPEKU11587
Supplier: MARKET SOURCE PART PROCESS
Frequently Bought Together