ContactUsLinkComponent
Mouse Inhibin Beta E (INHbE) High-sensitive ELISA Kit for detecting INHbE in serum, plasma, tissue homogenates, other biological flu ids. Species Reactivity: Mouse (Mus musculus). Detection Range: 1.56-100pg/ml. Sensitivity: 0.58pg/ml. Format: 96-Wel 1 * 96 Tests
  MSPPEKU11860
Mouse Inhibin Beta E (INHbE) High-sensitive ELISA Kit for detecting INHbE in serum, plasma, tissue homogenates, other biological flu ids. Species Reactivity: Mouse (Mus musculus). Detection Range: 1.56-100pg/ml. Sensitivity: 0.58pg/ml. Format: 96-Wel 1 * 96 Tests
  MSPPEKU11860