Mouse Interleukin 10 (IL10) Mini sample ELISA Kit for detecting IL10 in serum, plasma, tissue homogenates, cell lysates, cell cultur e supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 18.8-1,200pg/ml. Se 1 * 96 Tests
Catalog # MSPPEKU11163
Supplier: MARKET SOURCE PART PROCESS