Rat Oxidized Low Density Lipoprotein (OxLDL) ELISA Kit for detecting OxLDL in serum, plasma, other biological fluids. Species Reacti vity: Rat (Rattus norvegicus). Detection Range: 31.2-2,000pg/ml. Sensitivity: 13.9pg/ml. Format: 96-Well Plate. Assay 1 * 96 Tests
Catalog # MSPPEKU06446
Supplier: MARKET SOURCE PART PROCESS