Human Fc Fragment Of IgG Low Affinity IIIa Receptor (FcgR3A) ELISA Kit for detecting FcgR3A in serum, plasma, tissue homogenates, ce ll lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 31.2-2,000pg/ml. Sens 1 * 96 Tests
Catalog # MSPPEKU04063
Supplier: MARKET SOURCE PART PROCESS