Pig Leptin (LEP) ELISA Kit for detecting LEP in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biol ogical fluids. Species Reactivity: Pig (Sus scrofa Porcine). Detection Range: 0.312-20ng/ml. Sensitivity: 0.113ng/ml 1 * 96 Tests
Catalog # MSPPEKU05595
Supplier: MARKET SOURCE PART PROCESS
Frequently Bought Together