Mouse Interferon Gamma (IFNg) Mini sample ELISA Kit for detecting IFNg in serum, plasma, tissue homogenates, cell lysates, cell cult ure supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.6-1,000pg/ml. 1 * 96 Tests
Catalog # MSPPEKU11152
Supplier: MARKET SOURCE PART PROCESS