Human Pepsin (PP) ELISA Kit for detecting PP in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biol ogical fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 2.47-200ng/ml. Sensitivity: 0.87ng/ml. Form 1 * 96 Tests
Catalog # MSPPEKU08576
Supplier: MARKET SOURCE PART PROCESS
Frequently Bought Together