Deoxycholate (DC) ELISA Kit for detecting DC in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biol ogical fluids. Species Reactivity: Universal. Detection Range: 12.35-1,000ng/ml. Sensitivity: 4.91ng/ml. Format: 96-W 1 * 96 Tests
Catalog # MSPPEKU10789
Lieferant: MARKET SOURCE PART PROCESS
Frequently Bought Together