Human S100 Calcium Binding Protein A9 (S100A9) ELISA Kit for detecting S100A9 in serum, plasma, tissue homogenates, cell lysates, ce ll culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 15.6-1,000 1 * 96 Tests
Catalog # MSPPEKU07182
Supplier: MARKET SOURCE PART PROCESS