Human NLR Family, Pyrin Domain Containing Protein 3 (NLRP3) ELISA Kit for detecting NLRP3 in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. 1 * 96 Tests
Catalog # MSPPEKU10170
Supplier: MARKET SOURCE PART PROCESS