Human Eosinophil Peroxidase (EPX) ELISA Kit for detecting EPX in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 1.25-80ng/ml. Sensitivity: 0.51ng/ml. Format: 96-Well Plate. Assa 1 * 96 Tests
Catalog # MSPPEKU03918
Supplier: MARKET SOURCE PART PROCESS