Tryptophan (Trp) ELISA Kit for detecting Trp in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biol ogical fluids. Species Reactivity: Universal. Detection Range: 1.23-100ug/ml. Sensitivity: 0.55ug/ml. Format: 96-Well 1 * 96 Tests
Catalog # MSPPEKU11496
Supplier: MARKET SOURCE PART PROCESS
Frequently Bought Together