ContactUsLinkComponent
Human Klotho (KL) ELISA Kit for detecting KL in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biol ogical fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. For 1 * 96 Tests
  MSPPEKU05510
Human Klotho (KL) ELISA Kit for detecting KL in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biol ogical fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. For 1 * 96 Tests
  MSPPEKU05510
Frequently Bought Together