Human Angiotensin I Converting Enzyme 2 (ACE2) ELISA Kit for detecting ACE2 in serum, plasma, other biological fluids. Species React ivity: Human (Homo sapiens). Detection Range: 15.6-1,000pg/ml. Sensitivity: 5.5pg/ml. Format: 96-Well Plate. Assay Ty 1 * 96 Tests
Catalog # MSPPEKU02399
Supplier: MARKET SOURCE PART PROCESS