Mouse NLR Family, Pyrin Domain Containing Protein 3 (NLRP3) ELISA Kit for detecting NLRP3 in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.312-20ng/ml. 1 * 96 Tests
Catalog # MSPPEKU12250
Supplier: MARKET SOURCE PART PROCESS